Learn More
Grainger M-STRUTS Extension Gas Spring, Stainless Steel, 18-11/16 in. Extended Length, 51 lbf Force, 1 ea.
Catalog No. 502783219
Encompass_Preferred
This gas spring is a mechanical device designed to assist in the movement of objects such as windows, covers, and hatchbacks by utilizing compressed gas and liquid pressure. It features a rod and piston mechanism housed in a stainless steel cylinder, providing reliable force for opening or closing applications. This extension-only strut should be installed with the rod facing down to ensure optimal performance and longevity.
  • Force of 51 lbf
  • Constructed from durable stainless steel
  • Maximum operating temperature of 400 °F
  • Stroke length of 6 in.
  • Extended length of 18-11/16 in.
  • Compressed length of 12-11/16 in.
  • Thread size of M8x1.25
  • Fixed resistance type
  • End fittings not included
Catalog No. 50-278-3219 Supplier Grainger Supplier No. 19MT65
This gas spring is a mechanical device designed to assist in the movement of objects such as windows, covers, and hatchbacks by utilizing compressed gas and liquid pressure. It features a rod and piston mechanism housed in a stainless steel cylinder, providing reliable force for opening or closing applications. This extension-only strut should be installed with the rod facing down to ensure optimal performance and longevity.
  • Force of 51 lbf
  • Constructed from durable stainless steel
  • Maximum operating temperature of 400 °F
  • Stroke length of 6 in.
  • Extended length of 18-11/16 in.
  • Compressed length of 12-11/16 in.
  • Thread size of M8x1.25
  • Fixed resistance type
  • End fittings not included