Learn More
Grainger M-Struts® Extension Strut, Stainless Steel, 8 in. Stroke, 1 ea.
Catalog No. 502782810
Encompass_Preferred
This stainless steel mechanical extension strut is designed for reliable, maintenance-free motion control and dampening. Engineered to replace conventional gas struts, this unit functions without seals, oil, or gas, ensuring consistent performance in demanding environments, including extreme temperature ranges, caustic conditions, and dusty areas. It is suitable for applications such as lab equipment, cold chambers, and industrial hatches.
  • Constructed from durable stainless steel with a plain finish for high corrosion resistance
  • Operational in temperature ranges up to 400 °F (205 °C)
  • Features an 8 in. stroke length with 24-11/16 in. extended length and 16-11/16 in. compressed length
  • Equipped with M6x1.0 mounting threads
  • Ideal for use in medical, pharmaceutical, and agricultural settings
Catalog No. 50-278-2810 Supplier Grainger Supplier No. 19MT81
This stainless steel mechanical extension strut is designed for reliable, maintenance-free motion control and dampening. Engineered to replace conventional gas struts, this unit functions without seals, oil, or gas, ensuring consistent performance in demanding environments, including extreme temperature ranges, caustic conditions, and dusty areas. It is suitable for applications such as lab equipment, cold chambers, and industrial hatches.
  • Constructed from durable stainless steel with a plain finish for high corrosion resistance
  • Operational in temperature ranges up to 400 °F (205 °C)
  • Features an 8 in. stroke length with 24-11/16 in. extended length and 16-11/16 in. compressed length
  • Equipped with M6x1.0 mounting threads
  • Ideal for use in medical, pharmaceutical, and agricultural settings