Learn More
Grainger M-STRUTS Extension Strut, Stainless Steel, 24-13/16 in, 96 lbf, 1 ea
Catalog No. 502782798
Encompass_Preferred
This stainless steel extension strut utilizes compressed gas and liquid to provide reliable assistance in opening and closing applications. Designed for consistent force, it is suitable for use in diverse industrial and mechanical environments where controlled motion and dampening are required. The unit should be installed with the rod facing downward to maintain internal performance.
  • Extended length of 24 13/16 in.
  • Compressed length of 16 13/16 in.
  • Stroke length of 8 in.
  • Operating force of 96 lbf
  • Rod diameter of 0.55 in. and cylinder diameter of 1.1 in.
  • Constructed from stainless steel
  • Rod thread size M10 x 1.5
  • End fittings not included
  • Maximum operating temperature of 400 °F
Catalog No. 50-278-2798 Supplier Grainger Supplier No. 19MT85
This stainless steel extension strut utilizes compressed gas and liquid to provide reliable assistance in opening and closing applications. Designed for consistent force, it is suitable for use in diverse industrial and mechanical environments where controlled motion and dampening are required. The unit should be installed with the rod facing downward to maintain internal performance.
  • Extended length of 24 13/16 in.
  • Compressed length of 16 13/16 in.
  • Stroke length of 8 in.
  • Operating force of 96 lbf
  • Rod diameter of 0.55 in. and cylinder diameter of 1.1 in.
  • Constructed from stainless steel
  • Rod thread size M10 x 1.5
  • End fittings not included
  • Maximum operating temperature of 400 °F