Learn More
Fine Science Tools Graefe Forceps, Titanium, Straight, 10 cm
SDP
Catalog No. NC9712576
Encompass
Tip Shape: Straight/ Tips: Serrated/ Tip Dimensions: 1 x 0.9mm/ Alloy / Material: Titanium/ Length: 10cm
Catalog No. NC9712576 Supplier Fine Science Tools Supplier No. 1165010
Tip Shape: Straight/ Tips: Serrated/ Tip Dimensions: 1 x 0.9mm/ Alloy / Material: Titanium/ Length: 10cm