Learn More
Cole-Parmer Extech FL700 Waterproof Pocket Fluoride Tester
Supplier:  Cole-Parmer 9935930
Encompass
This portable, IP57-rated meter is designed for quick and accurate measurement of fluoride ions in drinking water and other aqueous samples. The sensing electrode, measuring electronics, and display are all contained in one easy-to-use package. Meter features auto power-off after 10 minutes and a low-battery indicator.
Catalog No. NC0934025