The Extech RPM33 is a combination contact and laser photo tachometer designed for accurate measurement of rotational speed (RPM), linear surface speed, and length. It offers both contact (2 to 20,000 rpm) and non-contact (2 to 99,999 rpm) measurement modes with high accuracy. This device provides versatile measurement capabilities for various industrial applications.
- Large 5 digit backlit LCD display
- Microprocessor based with quartz crystal oscillator to maintain high accuracy
- Stores and recalls 10 data sets in memory with four parameters (measurement, max, min and average)
- Provides wide RPM (photo and contact) and linear surface speed/length (contact) measurements
- Laser guided for greater distance non-contact measurements up to 500 mm (1.6 ft) away
- Includes a contact wheel for rpm or linear surface speed/length measurements, 9V battery, reflective tape, and a soft case.