Learn More
Cole-Parmer Extech SD800 Temperature/Humidity/CO2 Data Logger
Supplier:  Cole-Parmer 7000132
Encompass
Use these digital data loggers to monitor food, chemical, drug, and equipment storage areas, cleanrooms, incubators, and more! The easy-to-read LCD allows you to view current readings while recording. Set the flexible data logging interval to collect sample data from 5 seconds to 10 minutes. Data logging software stamps and stores date/time readings on an SD card in an Excel format for easy transfer to a PC. These data loggers can be programmed to set-off alarms at preset values providing warning when critical enviromental factors pose jeopardy to processes, goods, or people. The compact rugged design allows you to place these data loggers just about anywhere.
Catalog No. NC0648609