Learn More
Electron Microscopy Sciences Extended Reach Micro Ruler Only
SDP
Catalog No. 50996798
Encompass_Preferred
Precision etched, non-glare ruler made from surgical stainless steel, only 3 mm wide. The ruler is calibrated in 0.1 mm graduations, 10 mm total range. Stainless steel handle 230 mm long. Balanced and angled for the ultimate in flexibility able to reach into areas which are impossible for other measuring devices. Total weight of the ruler is 26 g. Applications include microscopy, chemistry and anywhere precise measurement in clean and very confined areas are of critical importance.
Catalog No. 50-996-798 Supplier Electron Microscopy Sciences Supplier No. 62136ER
Precision etched, non-glare ruler made from surgical stainless steel, only 3 mm wide. The ruler is calibrated in 0.1 mm graduations, 10 mm total range. Stainless steel handle 230 mm long. Balanced and angled for the ultimate in flexibility able to reach into areas which are impossible for other measuring devices. Total weight of the ruler is 26 g. Applications include microscopy, chemistry and anywhere precise measurement in clean and very confined areas are of critical importance.