Learn More
MSC Fibre-Metal Face Shield Window, Propionate, Clear, 9 3/4 in. x 19 in., 1 ea
Catalog No. 5000507096
Encompass
This face shield window is designed for use in industrial, manufacturing, construction, and laboratory settings where protection from dust, dirt, and debris is required. The clear, high-performance window is injection molded from propionate to provide superior durability and protection. It is preformed at the factory to ensure an exact fit and a perfect seal when used with compatible headgear.
  • Injection molded from durable propionate
  • Preformed to the exact curve of crown protectors for easy installation
  • Meets ANSI Z87.1+ and CSA Z94.3 standards
  • Measures 19 in. wide by 9 3/4 in. long with a 0.060 in. thickness
  • Compatible with F400 and F500 faceshield headgear
  • Can be worn over most prescription eyewear
Catalog No. 50-005-07096 Supplier MSC Supplier No. 92143940
This face shield window is designed for use in industrial, manufacturing, construction, and laboratory settings where protection from dust, dirt, and debris is required. The clear, high-performance window is injection molded from propionate to provide superior durability and protection. It is preformed at the factory to ensure an exact fit and a perfect seal when used with compatible headgear.
  • Injection molded from durable propionate
  • Preformed to the exact curve of crown protectors for easy installation
  • Meets ANSI Z87.1+ and CSA Z94.3 standards
  • Measures 19 in. wide by 9 3/4 in. long with a 0.060 in. thickness
  • Compatible with F400 and F500 faceshield headgear
  • Can be worn over most prescription eyewear