Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mopec Extended Warranty, Large Maestro, Size - 73 in. or Larger
SDP

Catalog No. 22021880 Shop All Mopec Products
Catalog No. 22-021-880 Supplier Mopec Supplier No. SV113
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Extend your current warranty to reduce the risk of equipment downtime caused by defects or repairs.

Extended Warranty

  • Availability: Purchased at point of sale or During Standard warranty
  • Definition: Purchased extension of the original warranty, continuing coverage for defects
  • Cost: Paid (one-time-fee or bundled into purchase)
  • Provider: Manufacturer, authorized provider
  • Duration: Extended term (e.g., additional 1 to 3 years)
  • Coverage Scope: Same as standard warranty (defect-related only)
  • Parts Coverage: Only defective OEM parts
  • Labor and Travel: Included for issues requiring on-site repairs (labor, travel, diagnostics)
  • Preventative Maintenance: Included at the end of the Standard Warranty and 1 PM at the end of each remaining year of the Extended Warranty
  • Response Time/SLAs: May have limited priority
  • Customer Support Access: Full access to technical support
  • Best For: Extended peace of mind with OEM-only issues
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.