Learn More
Ace Glass, Inc. 1/2 D x 5 L Titanium Ultrasonic Extender
SDP
Catalog No. 50977120
Encompass
1/2 D x 5 L Titanium Ultrasonic Extender screws into the threaded end of a sonic horn. Extender has non-replaceable solid tips.
Catalog No. 50-977-120 Supplier Ace Glass, Inc. Supplier No. 981606
1/2 D x 5 L Titanium Ultrasonic Extender screws into the threaded end of a sonic horn. Extender has non-replaceable solid tips.