Learn More
Ace Glass, Inc. 1 D x 5 L Titanium Ultrasonic Extender
SDP
Catalog No. 50977121
Encompass
1 D x 5 L Titanium Ultrasonic Extender screws into the threaded end of a sonic horn. Extender has non-replaceable solid tips.
Catalog No. 50-977-121 Supplier Ace Glass, Inc. Supplier No. 981610
1 D x 5 L Titanium Ultrasonic Extender screws into the threaded end of a sonic horn. Extender has non-replaceable solid tips.