Learn More
Ace Glass, Inc. 3/4 D x 5 L Titanium Ultrasonic Extender
SDP
Catalog No. 50980545
Encompass
3/4 D x 5 L Titanium Ultrasonic Extender screws into the threaded end of a sonic horn. Extender has non-replaceable solid tips.
Catalog No. 50-980-545 Supplier Ace Glass, Inc. Supplier No. 981608
3/4 D x 5 L Titanium Ultrasonic Extender screws into the threaded end of a sonic horn. Extender has non-replaceable solid tips.