Learn More
Grainger Eaton Illuminated Push Button Operator, Extended, Clear, 30 mm, 120 V, 1 ea.
Catalog No. 502771926
Encompass_Preferred
The HT800 series momentary illuminated pushbutton operator provides reliable control for diverse industrial applications. Designed for 30 mm mounting, this non-metallic device features an extended operator head and transparent housing for clear contact block visibility. It is engineered with robust sealing technology to ensure performance in harsh environments.
  • Momentary operating mode with 120 V AC/DC illumination
  • Extended plastic operator with clear lens
  • Corrosion-resistant construction rated NEMA 3, 3R, 4, 4X, 12, and 13
  • Features gold-plated contacts for low-voltage reliability
  • Includes double-V gaskets and O-ring seals for watertight protection
  • Compatible with standard 30 mm panel cutouts
  • Suitable for industrial use with 6,000 operations per hour capacity
Catalog No. 50-277-1926 Supplier Grainger Supplier No. 13Z603
The HT800 series momentary illuminated pushbutton operator provides reliable control for diverse industrial applications. Designed for 30 mm mounting, this non-metallic device features an extended operator head and transparent housing for clear contact block visibility. It is engineered with robust sealing technology to ensure performance in harsh environments.
  • Momentary operating mode with 120 V AC/DC illumination
  • Extended plastic operator with clear lens
  • Corrosion-resistant construction rated NEMA 3, 3R, 4, 4X, 12, and 13
  • Features gold-plated contacts for low-voltage reliability
  • Includes double-V gaskets and O-ring seals for watertight protection
  • Compatible with standard 30 mm panel cutouts
  • Suitable for industrial use with 6,000 operations per hour capacity