Learn More
Arkray ExtendSURE Control Solution

Description
- Designed for Quality Assurance: Arkray ExtendSURE Control Solution verifies the accuracy and performance of compatible glucose meters and test strips, supporting consistent and reliable testing.
- Ensures Testing Confidence: Provides a dependable reference solution to confirm meter and strip functionality, helping users identify potential issues before they impact results.
- Supports Routine Quality Control: Ideal for daily, weekly, or periodic testing checks to maintain compliance with internal quality protocols.
- Easy-to-Use Formula: Simple application allows quick verification without complex steps or additional equipment.
- Helps Detect Meter or Strip Errors: Useful for troubleshooting unexpected readings or ensuring the system is functioning correctly after storage changes or new strip lots.
- Consistent Performance: Manufactured to provide stable, reproducible results across multiple tests to support accuracy monitoring.
- Compatible With Arkray Glucose Testing Systems: Formulated to work seamlessly with designated Arkray glucose meters and test strips (refer to product manual for exact compatibility).
- Ideal for Clinical and Home Use: Suitable for hospitals, clinics, and home users who require dependable glucose system performance validation.
- Supports Compliance: Helps facilities meet quality control requirements outlined by regulatory bodies and internal standards.
- Improves Long-Term Reliability: Routine use of Arkray ExtendSURE Control Solution helps maintain optimal meter performance and ensures accurate reporting for diabetes management.
Specifications
Specifications
| Sample Type | Urine |
| Type | Solution |
| Form | Liquid |
For quality control verification only. Not intended for patient testing.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.