Learn More
Waters Corp Waters Corporation Extension Arm Kit, for CH-A Column Heater, MS Interface
Catalog No. 50791695 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This extension arm kit is designed to facilitate the interface between a CH-A Column Heater and a Mass Spectrometry (MS) source. It ensures proper positioning and includes essential components for a complete setup.
  • Designed to extend the CH-A column heater for compatibility with an MS source.
  • Includes a mounting bracket for secure installation.
  • Features drip trays to manage potential spills or condensation.
  • Required for optimal interfacing of the column heater with mass spectrometry applications.
Catalog No. 50-791-695 Supplier Waters Corp Supplier No. 205000726
This extension arm kit is designed to facilitate the interface between a CH-A Column Heater and a Mass Spectrometry (MS) source. It ensures proper positioning and includes essential components for a complete setup.
  • Designed to extend the CH-A column heater for compatibility with an MS source.
  • Includes a mounting bracket for secure installation.
  • Features drip trays to manage potential spills or condensation.
  • Required for optimal interfacing of the column heater with mass spectrometry applications.