Learn More
Perkin Elmer US LLC Extension Autozero Cylinder
Catalog No. 502113438
Encompass_Preferred
The Extension Autozero Cylinder is a reference standard used to calibrate the TMA 7, DMA 7, and DMA 7e analyzers. It comes standard in the TMA 7 Extension Analysis Kit (N5190619).
Catalog No. 50-211-3438 Supplier Perkin Elmer US LLC Supplier No. N5191792
The Extension Autozero Cylinder is a reference standard used to calibrate the TMA 7, DMA 7, and DMA 7e analyzers. It comes standard in the TMA 7 Extension Analysis Kit (N5190619).