Learn More
Grainger Snap-Loc Extension Bar Set, Industrial Steel, 15 in. Length, 1 ea.
Catalog No. 502783775
Encompass_Preferred
This extension bar set is designed to work with the dolly system to extend the height of push bars and panel bars, enabling the safer and easier transport of taller cargo items. These industrial-grade steel bars facilitate the transformation of two push bars into a panel bar set, providing containment for large items such as lumber and pipe.
  • Constructed from 1-1/4 in. heavy-duty industrial steel with a weather-resistant, powder-coated finish
  • Includes scratch and corrosion-resistant, zinc-plated steel connectors
  • Features quick-release pins for fast connection and disconnection
  • Measures 38.1 cm (15 in.) in length
  • Rated for a 400 lb. load capacity
Catalog No. 50-278-3775 Supplier Grainger Supplier No. 19H369
This extension bar set is designed to work with the dolly system to extend the height of push bars and panel bars, enabling the safer and easier transport of taller cargo items. These industrial-grade steel bars facilitate the transformation of two push bars into a panel bar set, providing containment for large items such as lumber and pipe.
  • Constructed from 1-1/4 in. heavy-duty industrial steel with a weather-resistant, powder-coated finish
  • Includes scratch and corrosion-resistant, zinc-plated steel connectors
  • Features quick-release pins for fast connection and disconnection
  • Measures 38.1 cm (15 in.) in length
  • Rated for a 400 lb. load capacity