Learn More
Heidolph™ Extension cable
SDP
Catalog No. 036210769 Shop All Heidolph North America Products
Change view
Click to view available options
Includes:
Cable
Catalog No. Includes
03-621-0769 Cable
1 options
Catalog No. 03-621-0769 Supplier Heidolph™ Supplier No. 036210769
Add to Cart
Add to Cart
Allows quick and easy operation even outside closed fume hoods
Accessory for use with all Hei-VAP Rotary Evaporator Models. 1.5m Extension Cable to allow for quick and easy operation even outside of closed fume hoods
Specifications
Type Rotary Evaporator Accessory
For Use With (Equipment) Hei-VAP Core, Hei-VAP Expert, Hei-VAP Ultimate, Hei-VAP Expert, Hei-VAP Expert Control, Hei-VAP Ultimate Control Rotary Evaporator
Includes Cable
Length (Metric) 1.5 m
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov