Learn More
Julabo USA Inc Each, 3.5m extention cable for Pt100 sensors
SDP
Catalog No. 507534128
Encompass_Preferred
Bath Accessories, Julabo, 3.5 m Extension cable for Pt100 sensor, Lemosa-type. Highly recommended for use with PRESTO W91, W91tt, W91x, W91ttx, W92, W92tt, W92x, W92ttx.
Catalog No. 50-753-4128 Supplier Julabo USA Inc Supplier No. 8981103
Bath Accessories, Julabo, 3.5 m Extension cable for Pt100 sensor, Lemosa-type. Highly recommended for use with PRESTO W91, W91tt, W91x, W91ttx, W92, W92tt, W92x, W92ttx.