Learn More
Chemglass Life Sciences Aluminum Extension Plate
SDP
Catalog No. CG199509
Encompass_Preferred
Extension Plate. Aluminum extension plate fits on top of all 135mm diameter circular top hot plate stirrers. Increases the surface area from 135mm to 200mm.
Catalog No. CG199509 Supplier Chemglass Life Sciences Supplier No. CG199509
Extension Plate. Aluminum extension plate fits on top of all 135mm diameter circular top hot plate stirrers. Increases the surface area from 135mm to 200mm.