This extension pole and hook system is designed for ground-level installation of overhead fall arrest anchorages, allowing workers to establish secure connection points before ascending. By enabling the placement of a hook anchor onto overhead structures from the ground, the system protects the first climber prior to reaching height. The assembly features a lightweight fiberglass pole and a spring-loaded hook, capable of reaching structures up to 20 ft. (6 m) above ground level.
- Provides a secure anchor point for fall arrest systems
- Installs to angle iron, piping, and related structural members
- Constructed with a lightweight fiberglass pole
- Features a spring-loaded hook for reliable connection
- Reaches up to 20 ft. (6 m) from ground level
- Supports a capacity of 400 lbs.
- Meets applicable OSHA, ANSI, and CSA standards