Learn More
MSC Mitutoyo Extension Rod, Steel, 2 in., 1 ea.
Catalog No. 5000515124
Encompass
This extension rod provides a length of 50 mm (2 in.) and is designed for use with imperial indicators and series 7 depth gauges.
  • Manufactured from durable steel
  • Features a 4-48 UNF thread
  • Includes a tip
Catalog No. 50-005-15124 Supplier MSC Supplier No. 92116672
This extension rod provides a length of 50 mm (2 in.) and is designed for use with imperial indicators and series 7 depth gauges.
  • Manufactured from durable steel
  • Features a 4-48 UNF thread
  • Includes a tip