Learn More
Hanna Pharmaceutical Supply Co., Inc. Straight-Type Extension Set, Standard Bore, Fixed Collar, 34""
Catalog No. NC1157035
Encompass_Preferred
Extension Set with Male Luer Lock Adapter. Approximate Volume 3.8 mL. Approximate Length 34" (86 cm). Contains DEHP. Sterile Package, Nonpyrogenic. 50/CS
Please note: This item is nonrefundable, non-returnable and non-cancelable once order is placed.
This item is not returnable. View return policy
Catalog No. NC1157035 Supplier Hanna Pharmaceutical Supply Co., Inc. Supplier No. BAX2C6227
Extension Set with Male Luer Lock Adapter. Approximate Volume 3.8 mL. Approximate Length 34" (86 cm). Contains DEHP. Sterile Package, Nonpyrogenic. 50/CS
Please note: This item is nonrefundable, non-returnable and non-cancelable once order is placed.