Learn More
Hanna Pharmaceutical Supply Co., Inc. Straight-Type Extension Set
Catalog No. NC0047248
Encompass_Preferred
Extension Set MICRO-VOLUME with Male Luer Lock Adapter. Approximate Volume 0.32 mL. Approximate Length 36" (91 cm). Non-DEHP. Sterile Package, Nonpyrogenic. 50/CS
Please note: This item is nonrefundable, non-returnable and non-cancelable once order is placed.
This item is not returnable. View return policy
Catalog No. NC0047248 Supplier Hanna Pharmaceutical Supply Co., Inc. Supplier No. BAX2N3345
Extension Set MICRO-VOLUME with Male Luer Lock Adapter. Approximate Volume 0.32 mL. Approximate Length 36" (91 cm). Non-DEHP. Sterile Package, Nonpyrogenic. 50/CS
Please note: This item is nonrefundable, non-returnable and non-cancelable once order is placed.