Learn More
Hanna Pharmaceutical Supply Co., Inc. Straight-Type Catheter Extension Set
Catalog No. NC0304138
Encompass_Preferred
Straight-Type Catheter Extension Set Microbore and Male Luer Lock Adapter. Approximate Volume 0.26 mL. Approximate Length 6.8" (17 cm). Non-DEHP. Sterile Package, Nonpyrogenic. 50/CS
Please note: This item is nonrefundable, non-returnable and non-cancelable once order is placed.
This item is not returnable. View return policy
Catalog No. NC0304138 Supplier Hanna Pharmaceutical Supply Co., Inc. Supplier No. BAX2N1194
Straight-Type Catheter Extension Set Microbore and Male Luer Lock Adapter. Approximate Volume 0.26 mL. Approximate Length 6.8" (17 cm). Non-DEHP. Sterile Package, Nonpyrogenic. 50/CS
Please note: This item is nonrefundable, non-returnable and non-cancelable once order is placed.