Learn More
BURKLE INC Extension V2A, 39.37 in. (100 cm), SiloDrill
Catalog No. 5000026086
Encompass_Preferred
With the SiloDrill extension, the SiloDrill can be extended by one meter to a maximum of 350 cm. This means that bulk materials can also be sampled from deep silos.
Catalog No. 50-000-26086 Supplier BURKLE INC Supplier No. 53010500
With the SiloDrill extension, the SiloDrill can be extended by one meter to a maximum of 350 cm. This means that bulk materials can also be sampled from deep silos.