Learn More
Waters Corp External Cable, D-Type, 2.5 ft
Catalog No. 50791991 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Replacement control cable assembly, D26 to D26 Male to Female, 2.5ft. This spare part is used on the ACQUITY UPLC System from the Sample Manager to the High Temperature Column Heater. It is also used on the Preparative Chromatography Rack for the 25X5 series of pumps.
Catalog No. 50-791-991 Supplier Waters Corp Supplier No. 441000871
Replacement control cable assembly, D26 to D26 Male to Female, 2.5ft. This spare part is used on the ACQUITY UPLC System from the Sample Manager to the High Temperature Column Heater. It is also used on the Preparative Chromatography Rack for the 25X5 series of pumps.