Learn More
InBios International, Inc. External Controls - COVID-19/Flu A and B RespiraDX™ Rapid Test

Description
- Contains positive and negative controls
- Store refrigerated or at room temperature
- For in vitro diagnostic use
Specifications
Specifications
| CE Marker | No |
| Clia Complexity | Waived |
| Detectable Analytes | COVID-19 |
| DoA Calibrators | No |
| Type | Rapid Test |
| Disposable | Single-use |
| Form | Swab |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.