Learn More
Kuhner Shaker Inc EXTERNAL DIMENSIONS H771 X W1
Catalog No. 502569926
Encompass
ISF1-ZC Peltier with CO2 and HumidityISF1-Z with heating, shaking and Peltier humidity controlTemperature range: ambient -5C to 60CControl of 0.1C, Accuracy +/- 0.2C at 37CAccumid humidity control to 85%r.H. Set point 1%+/-2%r.HCO2 Control 0-20%, Set point 0.1% +/-0.02% (IR)One-tool multi-diameter hub (3, 6, 9, 12.5, 19, 25, 50, 70mm)DC direct beltless forever-drive and high load bearingsLoads up to 60kg (application dependent)100% stainless steel interiorExternal Dimensions: H771 x W1285 x D884mmWeight: 210kgFIVE YEAR WARRANTY HARDWARE,1 YEAR WARRANTY ON SENSORS
Catalog No. 50-256-9926 Supplier Kuhner Shaker Inc Supplier No. SMZ1503PC
Add to Cart
Add to Cart
Request a Quote
ISF1-ZC Peltier with CO2 and HumidityISF1-Z with heating, shaking and Peltier humidity controlTemperature range: ambient -5C to 60CControl of 0.1C, Accuracy +/- 0.2C at 37CAccumid humidity control to 85%r.H. Set point 1%+/-2%r.HCO2 Control 0-20%, Set point 0.1% +/-0.02% (IR)One-tool multi-diameter hub (3, 6, 9, 12.5, 19, 25, 50, 70mm)DC direct beltless forever-drive and high load bearingsLoads up to 60kg (application dependent)100% stainless steel interiorExternal Dimensions: H771 x W1285 x D884mmWeight: 210kgFIVE YEAR WARRANTY HARDWARE,1 YEAR WARRANTY ON SENSORS