Learn More
Med Vet International External Fixation Rod, J0371RLA, 5/16" x 4", Each
Catalog No. 5000581921
Encompass_Preferred
Carbon fiber ESF bars offer two advantages over conventional stainless. They are radiolucent and lighter in weight but comparable in strength. Note: carbon fiber needs to be cut with a saw versus stainless pin cutters.
Catalog No. 50-005-81921 Supplier Med Vet International Supplier No. JOR371RLA
Carbon fiber ESF bars offer two advantages over conventional stainless. They are radiolucent and lighter in weight but comparable in strength. Note: carbon fiber needs to be cut with a saw versus stainless pin cutters.