Learn More
Med Vet International External Fixation Rod, J0371RLB, 5/16" x 6", Each
Catalog No. 5000582033
Encompass_Preferred
Carbon fiber ESF bars offer two advantages over conventional stainless. They are radiolucent and lighter in weight but comparable in strength. Note: carbon fiber needs to cut with a saw versus stainless pin cutters.
Catalog No. 50-005-82033 Supplier Med Vet International Supplier No. JOR371RLB
Carbon fiber ESF bars offer two advantages over conventional stainless. They are radiolucent and lighter in weight but comparable in strength. Note: carbon fiber needs to cut with a saw versus stainless pin cutters.