Learn More
Med Vet International External Fixation Rod, J0371RSB, 1/8" x 2", Each
Catalog No. 5000582042
Encompass_Preferred
Carbon fiber ESF bars offer two advantages over conventional stainless. They are radiolucent and lighter in weight but comparable in strength. Note carbon fiber need to cut with a saw versus stainless pin cutters.
Catalog No. 50-005-82042 Supplier Med Vet International Supplier No. JOR371RSB
Carbon fiber ESF bars offer two advantages over conventional stainless. They are radiolucent and lighter in weight but comparable in strength. Note carbon fiber need to cut with a saw versus stainless pin cutters.