Learn More
Med Vet International External Fixation Rod, J0371RSF, 1/8" x 6", Each
Catalog No. 5000581922
Encompass_Preferred
Carbon fiber ESF bars offer two advantages over conventional stainless. They are radiolucent and lighter in weight but comparable in strength. Note carbon fiber need to cut with a saw versus stainless pin cutters.
Catalog No. 50-005-81922 Supplier Med Vet International Supplier No. JOR371RSF
Carbon fiber ESF bars offer two advantages over conventional stainless. They are radiolucent and lighter in weight but comparable in strength. Note carbon fiber need to cut with a saw versus stainless pin cutters.