Learn More
Azzota Corp External Na Lamp Abbe Refract
Catalog No. 5000887316
Encompass_Preferred
External Na Light Source for Abbe Refractometer Na Lamp at 589nm wavelength Height adjustable 110V or 220V
Catalog No. 50-008-87316 Supplier Azzota Corp Supplier No. ARLSNA
External Na Light Source for Abbe Refractometer Na Lamp at 589nm wavelength Height adjustable 110V or 220V