Learn More
METTLER TOLEDO™ External Relay for Intracooler
Catalog No. 01915200 Shop All METTLER TOLEDO Products
Change view
Click to view available options
For Use With (Equipment):
30319177, 30319178
Catalog No. For Use With (Equipment)
01-915-200 30319177, 30319178
1 options
This item is not returnable. View return policy
Catalog No. 01-915-200 Supplier METTLER TOLEDO™ Supplier No. 51143415
Add to Cart
Add to Cart
This item is not returnable. View return policy
External Relay for Intracooler
Automatically sends singals to turn the intracooler on or off depending on the power state of the DSC.
For use with 30319177 & 30319178
TRUSTED_SUSTAINABILITY
Specifications
Description External Relay
For Use With (Equipment) 30319177, 30319178