Learn More
Anton Paar External temperature sensor for Multicube 48
Catalog No. NC1818273
Encompass_Preferred
Temperature sensor to measure the solution temperature directly in the vial (includes holder).Suitable for use with Anton Paar Multicube 48 Hot block digestion heating unit NC1818247
Catalog No. NC1818273 Supplier Anton Paar Supplier No. 224678
Temperature sensor to measure the solution temperature directly in the vial (includes holder).Suitable for use with Anton Paar Multicube 48 Hot block digestion heating unit NC1818247