Learn More
Research Products International Corp CryoFreeze Externally Threaded Tubes with Caps, 1.8ml Capacity, 500 per Package
SDP
Catalog No. 501537661
Encompass
CryoFreeze Cryogenic Tubes are designed for safe storage and transportation of biological materials. Ideal for a variety of applications including boiling, freezing and centrifuging. Tubes are manufactured from FDA-approved, medical grade polypropylene with HDPE caps. Tubes feature pre-printed black 100μL graduations and a significant white marking area on all tubes. Self standing tubes offer stargrip interlocking base and allow one-hand use in workstations.
Tubes comply with the In Vitro Devices Directive (IVD). Packaged in resealable stand-up pouches that minimize the use of benchtop space.
Withstand -196°C (vapor)
FDA-approved, medical grade
Sterile, RNase, DNase-free
Clearly marked graduations
Catalog No. 50-153-7661 Supplier Research Products International Corp Supplier No. 212601
CryoFreeze Cryogenic Tubes are designed for safe storage and transportation of biological materials. Ideal for a variety of applications including boiling, freezing and centrifuging. Tubes are manufactured from FDA-approved, medical grade polypropylene with HDPE caps. Tubes feature pre-printed black 100μL graduations and a significant white marking area on all tubes. Self standing tubes offer stargrip interlocking base and allow one-hand use in workstations.
Tubes comply with the In Vitro Devices Directive (IVD). Packaged in resealable stand-up pouches that minimize the use of benchtop space.
Withstand -196°C (vapor)
FDA-approved, medical grade
Sterile, RNase, DNase-free
Clearly marked graduations