Learn More
Grainger Externalooving Blade KGDR 2T17C Ext
Catalog No. 502763831
Encompass_Preferred
The Kyocera 170HW4 is a high-speed, high-volume, wide-format black and white laser printer. It prints up to 170 pages per minute at 1200 x 1200 dpi resolution. The printer has a maximum paper capacity of 18,000 sheets and supports media sizes up to 13" x 19.2". Connectivity options include Ethernet, USB 2.0, and Parallel ports. Some key features include long-life consumables, instant-on fuser technology, and PCL6/PostScript 3 emulation. The 170HW4 is ideal for high-volume printing of technical documents, manuals, posters, and more.
Catalog No. 50-276-3831 Supplier Grainger Supplier No. 170HW4
The Kyocera 170HW4 is a high-speed, high-volume, wide-format black and white laser printer. It prints up to 170 pages per minute at 1200 x 1200 dpi resolution. The printer has a maximum paper capacity of 18,000 sheets and supports media sizes up to 13" x 19.2". Connectivity options include Ethernet, USB 2.0, and Parallel ports. Some key features include long-life consumables, instant-on fuser technology, and PCL6/PostScript 3 emulation. The 170HW4 is ideal for high-volume printing of technical documents, manuals, posters, and more.