Learn More
Grainger Externalooving Toolholder KGBAR 123
Catalog No. 502764164
Encompass_Preferred
The KYOCERA Indexable Parting and Grooving Insert, 170JA3, is a parting and grooving insert designed for use on steel, stainless steel, cast iron, and non-ferrous metals. It features a TiAlN PVD coating for increased wear resistance and heat resistance. The insert has a .118" thickness with .118" maximum depth of cut. It has an ISO SNGN geometry with a .197" inscribed circle and is compatible with ISO tool holders. The insert is made of micro-grain carbide for strength and durability.
Catalog No. 50-276-4164 Supplier Grainger Supplier No. 170JA3
The KYOCERA Indexable Parting and Grooving Insert, 170JA3, is a parting and grooving insert designed for use on steel, stainless steel, cast iron, and non-ferrous metals. It features a TiAlN PVD coating for increased wear resistance and heat resistance. The insert has a .118" thickness with .118" maximum depth of cut. It has an ISO SNGN geometry with a .197" inscribed circle and is compatible with ISO tool holders. The insert is made of micro-grain carbide for strength and durability.