Learn More
Grainger Externalooving Toolholder KGBAR 1641
Catalog No. 502764342
Encompass_Preferred
The Grainger website features indexable turning profiling toolholders for CNC lathes. These toolholders have a precision ground body to hold inserts for turning, facing, profiling and grooving operations. They are made from alloy steel or carbide and accept a variety of insert shapes including 80 degree diamond, 55 degree diamond, 35 degree diamond, and triangular. Toolholder sizes range from 0.5" to 1.5" and accept inserts with thicknesses between 0.08" and 0.25". The toolholders have a coolant-through option and clamping screw for secure insert retention. Overall, these toolholders provide rigid setups for repetitive turning operations.
Catalog No. 50-276-4342 Supplier Grainger Supplier No. 170JA8
The Grainger website features indexable turning profiling toolholders for CNC lathes. These toolholders have a precision ground body to hold inserts for turning, facing, profiling and grooving operations. They are made from alloy steel or carbide and accept a variety of insert shapes including 80 degree diamond, 55 degree diamond, 35 degree diamond, and triangular. Toolholder sizes range from 0.5" to 1.5" and accept inserts with thicknesses between 0.08" and 0.25". The toolholders have a coolant-through option and clamping screw for secure insert retention. Overall, these toolholders provide rigid setups for repetitive turning operations.