Learn More
Grainger Externalooving Toolholder KGBAR 1642
Catalog No. 502764412
Encompass_Preferred
The 170JA9 is a 9-inch bench grinder from JET Tools. It has a 1/2 HP, 1-phase, 115V motor that provides 3450 RPM. The grinding wheels are 9 inches in diameter, 1 inch wide, and have a 5/8 inch arbor size. The motor is protected with an overload switch to prevent overheating. The grinder has a steel base with rubber feet to reduce vibration. It comes with a transparent eye shield, spark deflector, and tool rests. This is a powerful, industrial-quality bench grinder suitable for sharpening tools and general purpose grinding.
Catalog No. 50-276-4412 Supplier Grainger Supplier No. 170JA9
The 170JA9 is a 9-inch bench grinder from JET Tools. It has a 1/2 HP, 1-phase, 115V motor that provides 3450 RPM. The grinding wheels are 9 inches in diameter, 1 inch wide, and have a 5/8 inch arbor size. The motor is protected with an overload switch to prevent overheating. The grinder has a steel base with rubber feet to reduce vibration. It comes with a transparent eye shield, spark deflector, and tool rests. This is a powerful, industrial-quality bench grinder suitable for sharpening tools and general purpose grinding.