Learn More
Grainger Externalooving Toolholder KGBAR 2525
Catalog No. 502764171
Encompass_Preferred
The Kyocera External Grooving Toolholder 170JD5 is a toolholder for external grooving and turning applications. It has a JIS D-type shank with a width of 1.5 inches and an overall length of 5 inches. The toolholder accepts grooving inserts with a thickness of 0.078-0.118 inches and a width of 0.078-0.197 inches. It is made of steel and has a black oxide finish. The 170JD5 is suitable for grooving, cutoff and profiling in most materials. It provides rigid clamping of inserts and allows high feed rates. This toolholder enables precise machining for various turning operations.
Catalog No. 50-276-4171 Supplier Grainger Supplier No. 170JD5
The Kyocera External Grooving Toolholder 170JD5 is a toolholder for external grooving and turning applications. It has a JIS D-type shank with a width of 1.5 inches and an overall length of 5 inches. The toolholder accepts grooving inserts with a thickness of 0.078-0.118 inches and a width of 0.078-0.197 inches. It is made of steel and has a black oxide finish. The 170JD5 is suitable for grooving, cutoff and profiling in most materials. It provides rigid clamping of inserts and allows high feed rates. This toolholder enables precise machining for various turning operations.