Learn More
Grainger Externalooving Toolholder KGBAL 1642
Catalog No. 502764234
Encompass_Preferred
The KGBAL-1642-170JC0 External Turning Toolholder from Kennametal is a right hand toolholder with a 1/2" shank diameter and a 1-1/2" x 1-1/2" insert size. It has a negative rake angle of -6 degrees and is made of steel for durability. This toolholder is designed for external turning applications and accepts CNMG style carbide inserts, providing rigidity and precision in roughing and finishing operations. With its secure clamping system, it allows for quick insert changes. The toolholder's precision ground pocket enables accurate insert positioning.
Catalog No. 50-276-4234 Supplier Grainger Supplier No. 170JC0
The KGBAL-1642-170JC0 External Turning Toolholder from Kennametal is a right hand toolholder with a 1/2" shank diameter and a 1-1/2" x 1-1/2" insert size. It has a negative rake angle of -6 degrees and is made of steel for durability. This toolholder is designed for external turning applications and accepts CNMG style carbide inserts, providing rigidity and precision in roughing and finishing operations. With its secure clamping system, it allows for quick insert changes. The toolholder's precision ground pocket enables accurate insert positioning.