Learn More
Grainger Externalooving Toolholder KGBAL 2020
Catalog No. 502764296
Encompass_Preferred
The KGBAL-2020-170JD3 is an external grooving toolholder from Kennametal. It has a shank diameter of 1 inch, an overall length of 5 inches, and accepts inserts with a thickness of 0.078-0.118 inches. The toolholder is made of steel and has a right hand orientation. It can be used for external grooving, profiling and cutoff applications. The toolholder has a maximum depth of cut of 0.315 inches.
Catalog No. 50-276-4296 Supplier Grainger Supplier No. 170JD3
The KGBAL-2020-170JD3 is an external grooving toolholder from Kennametal. It has a shank diameter of 1 inch, an overall length of 5 inches, and accepts inserts with a thickness of 0.078-0.118 inches. The toolholder is made of steel and has a right hand orientation. It can be used for external grooving, profiling and cutoff applications. The toolholder has a maximum depth of cut of 0.315 inches.