Learn More
Grainger Externalooving Toolholder KGBASL 202
Catalog No. 502764301
Encompass_Preferred
The Grainger 170JE2 is a Dayton 1/3 HP General Purpose Motor that runs on 115V AC power at 1725 RPM. It is a single phase, open drip proof motor with a 48Y frame and features sleeve bearings for quiet operation. This 1/3 HP motor can provide 0.25 horsepower and is thermally protected against overloads. With its compact size, it is well suited for driving pumps, fans, and shop tools requiring up to 1/3 horsepower.
Catalog No. 50-276-4301 Supplier Grainger Supplier No. 170JE2
The Grainger 170JE2 is a Dayton 1/3 HP General Purpose Motor that runs on 115V AC power at 1725 RPM. It is a single phase, open drip proof motor with a 48Y frame and features sleeve bearings for quiet operation. This 1/3 HP motor can provide 0.25 horsepower and is thermally protected against overloads. With its compact size, it is well suited for driving pumps, fans, and shop tools requiring up to 1/3 horsepower.