Learn More
Grainger Externalooving Toolholder KGBASL 202
Catalog No. 502763808
Encompass_Preferred
The External Grooving Toolholder KGBASL-202-170JC5 from Kennametal is a toolholder for external grooving and parting. It has a shank diameter of 0.7874 inches (20mm) and an overall length of 6.5 inches (165mm). The toolholder has an American National Standards Institute (ANSI) insert style of SCLCR/L with a maximum insert thickness of 0.118 inches (3mm). It is made of steel and can hold inserts with a thickness of 0.039 - 0.118 inches. The toolholder allows for easy indexing and precise adjustment.
Catalog No. 50-276-3808 Supplier Grainger Supplier No. 170JC5
The External Grooving Toolholder KGBASL-202-170JC5 from Kennametal is a toolholder for external grooving and parting. It has a shank diameter of 0.7874 inches (20mm) and an overall length of 6.5 inches (165mm). The toolholder has an American National Standards Institute (ANSI) insert style of SCLCR/L with a maximum insert thickness of 0.118 inches (3mm). It is made of steel and can hold inserts with a thickness of 0.039 - 0.118 inches. The toolholder allows for easy indexing and precise adjustment.