Learn More
Grainger Externalooving Toolholder KGDL 624JX
Catalog No. 502764198
Encompass_Preferred
The 170JV6 is a Dayton 1/3 HP, 115/230V AC, 1725 RPM, 56C Frame, Open Drip Proof, Ball Bearing, Sleeve Bearing, Electric Motor. It has a 1/3 HP rating, operates on 115/230 volt AC power, has a 1725 RPM speed, uses a 56C frame size, is open drip proof, utilizes ball and sleeve bearings, and is designed for general purpose applications. The motor is thermally protected, reversible, and CE compliant. It can be used with pumps, blowers, conveyors, and other industrial equipment needing 1/3 HP power.
Catalog No. 50-276-4198 Supplier Grainger Supplier No. 170JV6
The 170JV6 is a Dayton 1/3 HP, 115/230V AC, 1725 RPM, 56C Frame, Open Drip Proof, Ball Bearing, Sleeve Bearing, Electric Motor. It has a 1/3 HP rating, operates on 115/230 volt AC power, has a 1725 RPM speed, uses a 56C frame size, is open drip proof, utilizes ball and sleeve bearings, and is designed for general purpose applications. The motor is thermally protected, reversible, and CE compliant. It can be used with pumps, blowers, conveyors, and other industrial equipment needing 1/3 HP power.