Learn More
Grainger Externalooving Toolholder KGDL 3232C
Catalog No. 502764438
Encompass_Preferred
The 170JL8 External Grooving Toolholder from Kennametal is a high-quality toolholder designed for external grooving operations. It features a KGDL-3232C insert style and accepts inserts with a .118" thickness. The toolholder has a 1/2" shank diameter and is made from steel for durability. With its precision ground pocket, it provides accurate seating and secure holding of inserts. This external grooving toolholder enables efficient grooving, parting off, and cutoff operations. Its innovative design delivers reliable performance and consistent results. The 170JL8 is ideal for use on CNC lathes and machining centers.
Catalog No. 50-276-4438 Supplier Grainger Supplier No. 170JL8
The 170JL8 External Grooving Toolholder from Kennametal is a high-quality toolholder designed for external grooving operations. It features a KGDL-3232C insert style and accepts inserts with a .118" thickness. The toolholder has a 1/2" shank diameter and is made from steel for durability. With its precision ground pocket, it provides accurate seating and secure holding of inserts. This external grooving toolholder enables efficient grooving, parting off, and cutoff operations. Its innovative design delivers reliable performance and consistent results. The 170JL8 is ideal for use on CNC lathes and machining centers.