Learn More
Grainger Externalooving Toolholder KGDL 123T2
Catalog No. 502763821
Encompass_Preferred
The Kyocera External Grooving Toolholder 170JG0 is a toolholder designed for external grooving operations. It has a shank diameter of 0.625 inches and an overall length of 4 inches. The toolholder has a maximum grooving depth capacity of 0.315 inches and accepts grooving tool bits with thicknesses from 0.079 to 0.197 inches. It is made of steel and has an ISO designation of PCLNR/L 2020K12. The toolholder allows for high precision grooving, threading and cutoff operations. Its precision ground pocket seats tools at the centerline for excellent repeatability.
Catalog No. 50-276-3821 Supplier Grainger Supplier No. 170JG0
The Kyocera External Grooving Toolholder 170JG0 is a toolholder designed for external grooving operations. It has a shank diameter of 0.625 inches and an overall length of 4 inches. The toolholder has a maximum grooving depth capacity of 0.315 inches and accepts grooving tool bits with thicknesses from 0.079 to 0.197 inches. It is made of steel and has an ISO designation of PCLNR/L 2020K12. The toolholder allows for high precision grooving, threading and cutoff operations. Its precision ground pocket seats tools at the centerline for excellent repeatability.